missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ 384-Well Polypropylene Storage Block

Product Code. 10266141 Shop All Corning Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility unitSize
10266141 Non-sterile Pack of 100
10554581 Sterile Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10266141 Supplier Corning™ Supplier No. 3965
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Rigid sidewalls for optimal performance in automated handling

Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI
Format Standard
Lid Without Lid
Array 16 x 24
Color Clear
Material Polypropylene
No. of Wells 384
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Round
For Use With (Application) For high throughput applications requiring added volume
No. per Case 100/Cs.
No. per Pack 5/Pk.
Quantity 100/Cs.
Well Color Clear
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.