missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ 384-Well Polypropylene Storage Block

Product Code. p-156353 Shop All Corning Products
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Pack of 25
This item is not returnable. View return policy

Product Code. 10554581

Brand: Corning™ 3964

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Rigid sidewalls for optimal performance in automated handling

Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI
Format Standard
Lid Without Lid
Array 16 x 24
Color Clear
Material Polypropylene
No. of Wells 384
Sterility Sterile
Surface Treatment Non-treated
Well Shape Round
For Use With (Application) For high throughput applications requiring added volume
No. per Case 25/Cs.
No. per Pack 5/Pk.
Quantity 25/Cs.
Well Color Clear
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.