missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cosibelimab Recombinant Monoclonal Antibody

Product Code. 30284791
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30284791 1 mg 1mg
30282637 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30284791 Supplier Invitrogen™ Supplier No. MA559198

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. Se returpolicy

Recombinant Monoclonal Antibody

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.

Cosibelimab is a fully-human monoclonal IgG1 antibody that blocks the interaction between PD-L1 and PD-1 and certain receptors called B7. 1 by binding to PD-L1.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Cosibelimab
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 25mM Histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Gene Alias CK-301
Host Species Human
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.