missing translation for 'onlineSavingsMsg'
Learn More

Ecolab COSA PUR 85 Cleaning Booster

Product Code. 17869864
Click to view available options
pH:
3 to 3.6
Unit Size:
21kg
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 17869864

Brand: Ecolab 2323210

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

COSA PUR 85 is a liquid oxidative cleaning booster for alkaline cleaning solutions in the pharmaceutical and cosmetic industries.

  • Especially suitable for the removal of residues which can only be digested or decomposed oxidatively.
  • Has oxidising properties without chlorine.
  • High soil-suspending power with special surfactants.
  • Foam-inhibited >40°C.
  • For general cleaning: Add COSA PUR 85 immediately before cleaning. Concentration: 1.0 to 2.0% COSA CIP solution, 0.5 to 1.0% COSA PUR 85, Temperature: 50°C to 85°C (circulating manner), Contact time: depends on degree and type of soiling.
  • COSA PUR 85 only shows its full effectiveness at a temperature >60°C by the separation of oxygen bubbles.
  • To achieve an optimum efficacy for a longer period, the product should be added in intervals.
  • Concentration, temperature and contact time can be optimized by evaluation of respective cleaning trials.
  • Final rinse with water of minimum drinking water quality, ensuring all soil and product residues are removed.
  • IMPORTANT - COSA PUR 85 is not intended to be used in closed systems, due to the formation of pressure increase which is resulting from the oxygen separation.

Specifications

Product Type Acid Cleaning Agent
Color-coded No
Form Concentrated Liquid
pH 3 to 3.6
Scent Unscented
Sterility Non-sterile
Appearance Clear
Autoclavable Not Autoclavable
Disposable Single-use
Filtered Filtered
Flammable No
Size 21 kg
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 21KG Cosa PUR 85 CIP additive Oxidising agent
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Hydrogen peroxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.