missing translation for 'onlineSavingsMsg'
Learn More

Ecolab COSA PUR 80 Surfactant

Product Code. 15796462
Change view
Click to view available options
pH:
9 to 10
Unit Size:
20kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH unitSize
15796462 9 to 10 20kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15796462 Supplier Ecolab Supplier No. 2323150

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

COSA PUR 80 is a sprayable, concentrated surfactant suitable for the removal of fatty production residues in the pharmaceutical and cosmetic industry.

  • Suitable for CIP-systems and manual cleaning.
  • High cleaning efficacy with fatty soils.
  • Suitable for addition to alkaline, neutral and acid application solutions.
  • For general CIP application (Pipelines, homogenizers, coaters, fermenters, etc): Concentration: 0.5 to 1.5% COSA PUR 80, Temperature: 60°C to 85°C, Contact time: depends on degree and type of soiling.
  • Before switching on the spray devices, it is recommended to heat up the solution to >50°C to prevent the formation of foam.
  • Concentration, temperature and contact time can be optimized by evaluation of respective cleaning trials.
  • For manual cleaning: Concentration: 0.5 to 1% COSA PUR 80, Temperature: 15°C to 50°C, Contact time: depends on degree and type of soiling. Use cleaning tools like swab, brush or pad if required.
  • Final rinse with water of minimum drinking water quality, ensuring all soil and product residues are removed.

Specifications

Product Type Mild Alkaline Liquid Detergent
Color-coded No
Form Concentrated Liquid
pH 9 to 10
Scent Unscented
Sterility Non-sterile
Appearance Clear Yellow
Autoclavable Not Autoclavable
Color Yellow
Disposable Single-use
Filtered Filtered
Flammable No
Size 20 kg
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • COSA PUR 80
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: alcohols C12-18, alkylamines ethoxylates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.