missing translation for 'onlineSavingsMsg'
Learn More

Ecolab COSA CIP 95 Liquid Detergent

Product Code. 15758049
Change view
Click to view available options
pH:
12.3-12.7
Unit Size:
26kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH unitSize
15758049 12.3-12.7 26kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15758049 Supplier Ecolab Supplier No. 2324940

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

COSA CIP 95 is a liquid alkaline detergent for the pharmaceutical , biotech and cosmetic industries.

  • Especially suitable for the removal of soiling like residues from tablet coverings in the pharmaceutical and cosmetic industry.
  • With the addition of special cleaning boosters,COSA CIP 95 is suitable for the cleaning of product lines with fatty and gel-containing residues.
  • Suitable for CIP-systems.
  • Highly efficient against residues from tablet coverings.
  • Surfactant free.
  • For general CIP application (Homogenizers, pipelines, coaters, tanks, etc): Concentration: 1 to 3% COSA CIP 95 solution, Temperature: 5°C to 95°C (ideally 60°C to 80°C), Contact time: depending on degree and type of soiling.
  • Concentration, temperature and cleaning time can be optimized by evaluation of respective cleaning trials.
  • Final rinse with water of minimum drinking water quality, ensuring all soil and product residues are removed.

Specifications

Product Type Alkaline Cleaning Agent
Color-coded No
Form Clear Liquid
pH 12.3-12.7
Scent Original
Sterility Non-sterile
Appearance Clear Brown
Autoclavable Not Autoclavable
Color Brown
Disposable Single-use
Filtered Filtered
Flammable No
Size 26 kg
Show More Show Less
compliance-icons
Product Identifier
  • COSA CIP 95
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/physician..
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.