Learn More
Isofield™ Cos Non-Sterile Lightweight Cleanroom Sock - GMP Annex 1 Compatible
GREENER_CHOICEBIOPHARMA
Product Code. p-200088580
Change view
Click to view available options
Size:
Large/X-Large
Small/Medium
Unit Size:
500 pairs
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
30178833 Small/Medium 500 pairs
30178852 Large/X-Large 500 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30178833 Supplier Isofield™ Supplier No. 70610SM
Only null left
Add to basket
Add to basket
The Isofield Cos single-use non-sterile lightweight cleanroom sock is GMP Annex 1 compatible, offering comfort and protection in cleanroom settings. Available in three sizes, the Cos’ hourglass heel design ensures comfort levels rivalling most reusable alternatives.
  • Non-sterile lightweight
  • Cleanroom socks
  • Single-use
  • 95% polyester, 5% spandex
  • Mid-calf length
  • Low linting
  • White color
Specifications
Size Small/Medium
Product Type Cleanroom Socks