missing translation for 'onlineSavingsMsg'
Learn More

Corynoxine, MedChemExpress

Product Code. 16449178
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16449178 10 mg 10mg
16441588 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16449178 Supplier Medchem Express Supplier No. HYN0901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Corynoxine, a tetracyclic oxindole alkaloid, is isolated from the hooks of Uncaria macrophylla. Corynoxine is a natural autophagy enhancer that promotes the clearance of alpha-synuclein via Akt/mTOR pathway.

Corynoxine, a tetracyclic oxindole alkaloid, is isolated from the hooks of Uncaria macrophylla. Corynoxine is a natural autophagy enhancer that promotes the clearance of alpha-synuclein via Akt/mTOR pathway.

Specifications

Chemical Name or Material Corynoxine
CAS 6877-32-3
Color White
Purity Grade Notes Research
Molecular Formula C22H28N2O4
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (260.10 mM)
SMILES O=C(NC1=C2C=CC=C1)[C@@]32[C@@](C[C@H](/C(C(OC)=O)=C\OC)[C@H](CC)C4)([H])N4CC3
Molecular Weight (g/mol) 384.47
Formula Weight 384.47
Percent Purity 99.58%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.