missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corydaline, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Corydaline ((+)-Corydaline), an isoquinoline alkaloid isolated from Corydalis yanhusuo, is an AChE inhibitor with an IC50 of 226 μM. Corydaline is a μ-opioid receptor (Ki of 1.23 μM) agonist and inhibits enterovirus 71 (EV71) replication (IC50 of 25.23 μM). Corydaline has anti-angiogenic, anti-allergic and gastric-emptying and antinociceptive activities.
Specifications
Specifications
| Chemical Name or Material | Corydaline |
| CAS | 518-69-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H27NO4 |
| Quantity | 10 mg |
| Synonym | (+)-Corydaline Corydalin |
| Solubility Information | DMSO : 33.33 mg/mL (90.22 mM; Need ultrasonic) |
| SMILES | C[C@@H](C1=CC=C2OC)[C@]3([H])C4=CC(OC)=C(OC)C=C4CCN3CC1=C2OC |
| Molecular Weight (g/mol) | 369.45 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction