missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cortisone, Lipomed
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Cortisone
Specifications
Specifications
| Chemical Name or Material | Cortisone |
| CAS | 53-06-5 |
| Analyte or Component Names | Cortisone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H28 O5 |
| Synonym | Hydrocortisone Imp. B (EP), Cortisone (8 CI), 17,21-Dihydroxypregn-4-ene-3,11,20-trione, 11-Dehydro-17-hydroxycorticosterone, 17-Hydroxy-11-dehydrocorticosterone, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | C[C@]12 CCC(=O)C=C1CC[C@H]3 [C@@H]4 CC[C@](O)(C(=O)CO)[C@@]4(C)CC(=O)[C@H]23 |
| IUPAC Name | (8 S,9 S,10 R,13 S,14 S,17 R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-1,2,6,7,8,9,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,11-dione |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction