missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cortisone 0.1 mg/ml in Methanol, LoGiCal
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Cortisone 0.1 mg/ml in Methanol
Specifications
Specifications
| Chemical Name or Material | Cortisone |
| CAS | 53-06-5 |
| Analyte or Component Names | Cortisone |
| Shipping Condition | Ice Pack (-20°C) |
| Molecular Formula | C21 H28 O5 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Hydrocortisone Imp. B (EP), Cortisone (8 CI), 17,21-Dihydroxypregn-4-ene-3,11,20-trione, 11-Dehydro-17-hydroxycorticosterone, 17-Hydroxy-11-dehydrocorticosterone, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione, Delta4-Pregnene-17 alpha,21-diol-3,11,20-trione |
| InChI Formula | InChI=1 S/C21H28O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h9,14-15,18,22,26 H,3-8,10-11H2,1-2H3/t14-,15-,18+,19-,20-,21-/m0/s1 |
| SMILES | C[C@]12 CCC(=O)C=C1CC[C@H]3 [C@@H]4 CC[C@](O)(C(=O)CO)[C@@]4(C)CC(=O)[C@H]23 |
| IUPAC Name | (8 S,9 S,10 R,13 S,14 S,17 R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-1,2,6,7,8,9,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,11-dione |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction