Learn More
Invitrogen™ Cortisol Monoclonal Antibody (F4P1A3)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA514676
Product Code. 11375133
  • £460.00 / 100µg
Description
MA5-14676 targets Cortisol in ELISA, IP, and RIA applications and shows reactivity with Chemical samples. The MA5-14676 immunogen is cortisol-3-CMO-BSA. Product MA514676 is a smaller package size of MIC0201 (formerly sold as a Seradyn product).
Cortisol is the most potent glucocorticoid produced by the human adrenal. It is synthesized from cholesterol and its production is stimulated by pituitary adrenocorticotropic hormone (ACTH) which is regulated by corticotropin releasing factor (CRF). ACTH and CRF secretions are inhibited by high cortisol levels in a negative feedback loop. In plasma a majority of cortisol is bound with high affinity to corticosteroid binding globulin (CBG or transcotin). Cortisol acts through specific intracellular receptors and affects numerous physiologic systems including immune function, glucose counter regulation, vascular tone, and bone metabolism.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cortisol
Monoclonal
1 mg/mL
0.01M potassium phosphate with 1 mg/mL BSA, 50% glycerol, 2.5 mM KCl, 0.135M NaCl and 0.05% sodium azide
Cortisol-3-CMO-BSA.
100 μg
Primary
Maintain refrigerated at 2°C to 8°C for up to 6 months. For long term storage store at -20°C
Liquid
Chemical
ELISA, Immunoprecipitation, Radioimmune Assays (RIA)
F4P1A3
Unconjugated
Mouse
Protein A
RUO
Chemical
Antibody
IgG2b