missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cortisol Monoclonal Antibody (CORT-1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA116703
This item is not returnable.
View return policy
Description
Cortisol Monoclonal Antibody for ELISA
Cortisol is the most potent glucocorticoid produced by the human adrenal. It is synthesized from cholesterol and its production is stimulated by pituitary adrenocorticotropic hormone (ACTH) which is regulated by corticotropin releasing factor (CRF). ACTH and CRF secretions are inhibited by high cortisol levels in a negative feedback loop. In plasma a majority of cortisol is bound with high affinity to corticosteroid binding globulin (CBG or transcotin). Cortisol acts through specific intracellular receptors and affects numerous physiologic systems including immune function, glucose counter regulation, vascular tone, and bone metabolism.
Specifications
| Cortisol | |
| Monoclonal | |
| 3.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Purified cortisol conjugated with BSA. | |
| 500 μg | |
| Primary | |
| 4°C, do not freeze | |
| Liquid | |
| Chemical |
| ELISA | |
| CORT-1 | |
| Unconjugated | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction