Learn More
Invitrogen™ Cortisol Monoclonal Antibody (BGN/C53)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA183090
Description
Mouse anti cortisol monoclonal antibody, clone BGN/C53 recognizes cortisol, a glucocorticoid hormone synthesized from cholesterol by the adrenal cortex. It is released in response to pituitary adenocorticotrophic hormone (ACTH) which is, in turn, regulated by corticotrophin releasing factor (CRF).
Cortisol is the most potent glucocorticoid produced by the human adrenal. It is synthesized from cholesterol and its production is stimulated by pituitary adrenocorticotropic hormone (ACTH) which is regulated by corticotropin releasing factor (CRF). ACTH and CRF secretions are inhibited by high cortisol levels in a negative feedback loop. In plasma a majority of cortisol is bound with high affinity to corticosteroid binding globulin (CBG or transcotin). Cortisol acts through specific intracellular receptors and affects numerous physiologic systems including immune function, glucose counter regulation, vascular tone, and bone metabolism.
Specifications
| Cortisol | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| Synthetic corticosterone conjugated to BSA. | |
| 200 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| Chemical |
| ELISA | |
| BGN/C53 | |
| Unconjugated | |
| Mouse | |
| Purified | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG3 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.