missing translation for 'onlineSavingsMsg'
Learn More

Cortisol, Lipomed

Product Code. 30463956
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30463956 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30463956 Supplier Lipomed Supplier No. LPMCOR163050
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Cortisol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydrocortisone
CAS 50-23-7
Analyte or Component Names Hydrocortisone
Shipping Condition Room Temperature
Molecular Formula C21 H30 O5
Synonym Hydrocortisone, Cortisol, 11 beta,17,21-Trihydroxy-pregn-4-ene-3,20-dione, Hydrocortisone Hydrogen Succinate Imp. A (EP), Prednisolone Imp. A (EP), 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione, 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES C[C@]12 CCC(=O)C=C1CC[C@H]3 [C@@H]4 CC[C@](O)(C(=O)CO)[C@@]4(C)C[C@H](O)[C@H]23
IUPAC Name (8 S,9 S,10 R,11 S,13 S,14 S,17 R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1 H-cyclopenta[a]phenanthren-3-one
Industry Type Pharmaceutical
Molecular Weight (g/mol) 362.46
Quantity 50 mg
Formula Weight 362.2093 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.