missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cortisol Antibody (BGN/C53), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62484AF594
This item is not returnable.
View return policy
Description
Cortisol Monoclonal specifically detects Cortisol in All Species samples. It is validated for ELISA.
Specifications
| Cortisol | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Synthetic corticosterone conjugated to Bovine Serum Albumin. | |
| Primary | |
| All species | |
| IgG3 |
| ELISA | |
| BGN/C53 | |
| ELISA | |
| Mouse | |
| 0.1 mL | |
| This product recognises the glucocorticoid hormone cortisol, produced in the adrenal cortex and involved in responses to stress. Cross reactivity: Cortisol 100%, Corticosterone 50%, Prednisolone 15%, 11-Desoxycortisol, <0.1% Cortisone,<0.1% 17-a-OH Progesterone, <0.1%. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction