missing translation for 'onlineSavingsMsg'
Learn More

Corticotropin Releasing Factor Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Product Code. 18388441
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18388441 20 μg 20µL
18056035 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18388441 Supplier Bio-Techne Supplier No. NBP31535620UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Corticotropin Releasing Factor Polyclonal antibody specifically detects Corticotropin Releasing Factor in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Corticotropin Releasing Factor
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000
Formulation PBS, 50% glycerol, pH7.3
Gene Alias corticoliberin, corticotropin releasing hormone, Corticotropin-releasing hormone, CRFCorticotropin-releasing factor
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 25-194 of human Corticotropin Releasing Factor (NP_000747.1). LLSRGPVPGARQAPQHPQPLDFFQPPPQSEQPQQPQARPVLLRMGEEYFLRLGNLNKSPAAPLSPASSLLAGGSGSRPSPEQATANFFRVLLQQLLLPRRSLDSPAALAERGARNALGGHQEAPERERRSEEPPISLDLTFHLLREVLEMARAEQLAQQAHSNRKLMEII
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Biology, Endocrinology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1392
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.