missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corticotropin Releasing Factor Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£141.00
Specifications
| Antigen | Corticotropin Releasing Factor |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Corticotropin Releasing Factor Polyclonal specifically detects Corticotropin Releasing Factor in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| Corticotropin Releasing Factor | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 1392 | |
| Antibody was raised against a 16 amino acid peptide near the amino terminus of human CRH. The immunogen is located within amino acids 30 - 80 of CRH. Amino Acid Squence: FQPPPQSEQPQQPQAR | |
| Primary | |
| CRH antibody is human specific. |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| corticoliberin, corticotropin releasing hormone, Corticotropin-releasing hormone, CRFCorticotropin-releasing factor | |
| CRH | |
| IgG | |
| Affinity Purified | |
| 29 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title