missing translation for 'onlineSavingsMsg'
Learn More

Abcam Corticosterone ELISA kit

Product Code. 32112035 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1 x 96 Tests
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32112035 1 x 96 Tests 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32112035 Supplier Abcam Supplier No. ab3237431x96Tests
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Corticosterone ELISA kit is a competitive ELISA designed to quantify mouse, rat and human corticosterone (CORT) in serum, plasma, saliva, urine, and milk with a sensitivity of 0.33 ng/mL.

Corticosterone ELISA kit is a competitive ELISA designed to quantify mouse, rat and human corticosterone (CORT) in serum, plasma, saliva, urine, and milk with a sensitivity of 0.33 ng/mL. - Wide dynamic range - quantifies 0.391-25 ng/mL - Fast assay time - complete results in 3 hours - High recovery rate - achieves 98% general recovery - New and improved version of ab108821 which was cited in over 150 publications

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Assay Range 0.391 to 25 ng/mL
Assay Sensitivity 0.33 ng/mL
Detectable Analytes Corticosterone ()
Assay Corticosterone ELISA kit
Immunoassay Kit Format Competitive ELISA Kit
Product Type Competitive ELISA
Sample Type Milk, Plasma, Saliva, Serum, Urine
Sufficient For 96 Tests
For Use With (Application) ELISA
Immunoassay Kit Grade RUO
Measuring Range 0.391 to 25 ng/mL
Quantitation Range 0.391 to 25 ng/mL
Quantity 1 x 96 Tests
Regulatory Status RUO
Shipping Condition Blue Ice
Target Corticosterone ()
Target Kit Named Corticosterone ()
Detection Limit 0.391 ng/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.