missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cortactin Recombinant Rabbit Monoclonal Antibody (23GB5130), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | Cortactin |
|---|---|
| Clone | 23GB5130 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087811
![]() |
Invitrogen™
MA552798 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088161
|
Invitrogen™
MA552799 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cortactin Recombinant Monoclonal antibody specifically detects Cortactin in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| Cortactin | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q14247, Q60598, Q66HL2 | |
| 13043, 2017, 60465 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5130 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 1110020L01Rik; amplaxin; cortactin; Cttn; Cttnb; EMS1; ems1 sequence (mammary tumor and squamous cell carcinoma-associated (p80/85 src substrate); FLJ34459; mammary tumor and squamous cell carcinoma associated (p80/85 src substrate); oncogene EMS1; Oncogene SRC8; Src substrate cortactin; src8; Unknown (protein for MGC:137923) | |
| A synthesized peptide derived from human Cortactin (30-84AA). | |
| Primary | |
| -20°C | |
| Cttn |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title