missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cortactin Recombinant Rabbit Monoclonal Antibody (2W5H9)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535798
This item is not returnable.
View return policy
Description
Immunogen sequence: MWKASAGHAV SIAQDDAGAD DWETDPDFVN DVSEKEQRWG AKTVQGSGHQ EHINIHKLRE NVFQEHQTLK EKELETGPKA SHGYGGKFGV EQDRMDKSAV.
Cortactin (Cttn) is a ubiquitous actin-binding protein that was originally identified as a substrate for Src. It contributes to the organization of the actin cytoskeleton and cell shape. Cortactin also plays a role in the formation of lamellipodia and cell migration. It is accumulated in peripheral, actin-enriched structures of cells suggesting that cortactin facilitates actin network formation. Cortactin has four major domains of interest the N-terminal acidic (NTA) and tandem repeats domains and the C-terminal proline-rich and SH3 Domains. NTA associates with the Arp2/3 and WASP complex at F-actin branches. Cortactin is involved in promoting cell motility and invasion, including a critical role in invadopodia, actin rich-subcellular protrusions associated with degradation of the ECM by cancer cells. Cortactin is phosphorylated by src family kinases at Y421, Y466, and Y482 and S405 and S418 that are phosphorylated by Erk family kinases.
Specifications
| Cortactin | |
| Recombinant Monoclonal | |
| 0.25 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14247, Q60598, Q66HL2 | |
| Cttn | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Cortactin (Q14247). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 2W5H9 | |
| Unconjugated | |
| Cttn | |
| 1110020L01Rik; amplaxin; cortactin; Cttn; Cttnb; EMS1; ems1 sequence (mammary tumor and squamous cell carcinoma-associated (p80/85 src substrate); FLJ34459; mammary tumor and squamous cell carcinoma associated (p80/85 src substrate); oncogene EMS1; Oncogene SRC8; Src substrate cortactin; src8; Unknown (protein for MGC:137923) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13043, 2017, 60465 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction