missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CORT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544997
This item is not returnable.
View return policy
Description
Peptide sequence: MPLSPGLLLL LLSGATATAA LPLEGGPTGR DSEHMQEAAG IRKSSLLTFL Sequence homology: Human: 100%.
This gene encodes a neuropeptide that is structurally similar to somatostatin. It binds to all known somatostatin receptors, and shares many pharmacological and functional properties with somatostatin, including the depression of neuronal activity. However, it also has many properties distinct from somatostatin, such as induction of slow-wave sleep, apparently by antagonism of the excitatory effects of acetylcholine on the cortex, reduction of locomotor activity, and activation of cation selective currents not responsive to somatostatin. The preproprotein undergoes further processing into multiple mature products. Read-through transcripts exist between this gene and the upstream APITD1 (apoptosis-inducing, TAF9-like domain 1) gene, as represented in GeneID:100526739.
Specifications
| CORT | |
| Polyclonal | |
| Unconjugated | |
| CORT | |
| CORT; Cortistatin; cortistatin-14; cortistatin-17; Cortistatin-29; CST-14; CST-17; CST-29; preprocortistatin; prepro-cortistatin; UNQ307/PRO350 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1325 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O00230 | |
| CORT | |
| Synthetic peptide directed towards the N-terminal of human CORT (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction