missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Coronin 1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546001
This item is not returnable.
View return policy
Description
Peptide sequence: STTTAADATP SGSLARAGEA GKLEEVMQEL RALRALVKEQ GDRICRLEEQ Sequence homology: Cow: 79%; Dog: 86%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 86%; Rabbit: 86%; Rat: 86%.
Members of the coronin family, such as CORO1B, are WD repeat-containing actin-binding proteins that regulate cell motility.
Specifications
| Coronin 1B | |
| Polyclonal | |
| Unconjugated | |
| Coro1b | |
| cor-1; Coro1b; coronin 1b; coronin 2; coronin, actin binding protein 1B; coronin, actin binding protein, 1B; coronin, actin binding protein, 1B (coronin 2); coronin, actin-binding protein, 1B; coronin-1B; coronin-1B-like; CORONIN-2; Coronin-like protein pp66; coroninse | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 57175 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BR76 | |
| Coro1b | |
| Synthetic peptide directed towards the C-terminal of human CORO1B (aa 431-480). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction