missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Coronin 1A Polyclonal Antibody
GREENER_CHOICE

Product Code. 11376153
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376153 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11376153 Supplier Invitrogen™ Supplier No. PA518186

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. Alternative splicing results in multiple transcript variants. A related pseudogene has been defined on chromosome 16.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Coronin 1A
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene CORO1A
Gene Accession No. P31146
Gene Alias actin-binding; bovine p57; Clabp; Clabp coronin; CLIPINA; Clipin-A; CORO1; CORO1A; CORO1A protein; coronin 1; coronin 1A; coronin, actin binding protein 1A; coronin, actin binding protein, 1A; coronin-1; coronin-1A; coronin-like actin binding protein; coronin-like protein A; Coronin-like protein p57; FLJ41407; HCORO1; IMD8; Lmb3; MGC117380; p57; TACO; tryptophan aspartate-containing coat protein
Gene Symbols CORO1A
Host Species Goat
Immunogen Synthetic peptide sequence (KRLDRLEETVQAK) corresponding to the C-terminus amino acids of CORO1A (aa 449-461).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11151
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.