missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coronin-1a Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB110-58867SS
This item is not returnable.
View return policy
Description
Coronin-1a Polyclonal specifically detects Coronin-1a in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Coronin-1a | |
| Polyclonal | |
| Unconjugated | |
| Supplied as serum with 5mM Sodium Azide | |
| clipin-A, CLIPINA, CORO1, coronin, actin binding protein, 1A, coronin, actin-binding protein, 1A, coronin-1, coronin-1A, Coronin-like protein A, Coronin-like protein p57, FLJ41407, HCORO1, MGC117380, p57, TACOCLABP, Tryptophan aspartate-containing coat protein | |
| Rabbit | |
| 55 kDa | |
| 0.025 mL | |
| Cell Cycle and Replication, Immunology, Innate Immunity, Neuroscience | |
| 11151 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1 mg/ml | |
| Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:500-1:1000 | |
| P31146 | |
| CORO1A | |
| C-terminal peptide of human coronin 1a coupled to KLH. [UniProt# P31146] | |
| Unpurified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction