missing translation for 'onlineSavingsMsg'
Learn More

Coronene, TRC

Product Code. 30356123
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30356123 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30356123 Supplier Toronto Research Chemicals Supplier No. TRCC695500100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Coronene
CAS 191-07-1
Molecular Formula C24 H12
Quantity 100 mg
Synonym Coronene, Circumbenzene, NSC 90725
InChI Formula InChI=1S/C24H12/c1-2-14-5-6-16-9-11-18-12-10-17-8-7-15-4-3-13(1)19-20(14)22(16)24(18)23(17)21(15)19/h1-12H
SMILES c1cc2ccc3ccc4ccc5ccc6ccc1c7c6c5c4c3c27
IUPAC Name coronene
Molecular Weight (g/mol) 300.35
Formula Weight 300.0939
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.