missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Coronavirus Spike Protein Monoclonal Antibody (07)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA529970
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human coronavirus spike glycoprotein.
Human coronaviruses are the major cause of upper respiratory tract illness, such as the common cold, in humans. Coronaviruses are positive-stranded RNA viruses, featuring the largest viral RNA genomes known to date (27-31 kb).
Specifications
| Coronavirus Spike Protein | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| 07 | |
| Unconjugated | |
| Cov2; Cov-2; Cov-2 Spike protein; COVID 19; Covid19 | |
| Recombinant Human coronavirus spike glycoprotein protein. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction