Learn More
CORO2A Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10119-1-AP-20UL
Description
The CORO2A antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CORO2A. This antibody recognizes human, mouse antigen. The CORO2A antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| CORO2A | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 7464 | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| 333 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |