Learn More
CORO2A Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10119-1-AP-150UL
 View more versions of this product
Product Code. 16836293
  • £349.00 / 150µL
Description
The CORO2A antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CORO2A. This antibody recognizes human, mouse antigen. The CORO2A antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
CORO2A
Polyclonal
Unconjugated
150 μL
7464
IgG
Western Blot, Immunohistochemistry, ELISA
333 μg/mL
Rabbit
Primary
Human, Mouse