Learn More
CORO1C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14749-1-AP-150UL
 View more versions of this product
Product Code. 16833554
  • £349.00 / 150µL
Description
Coronin 3 is associated with the aggression and metastasis of hepatocellular carcinoma. Coronins are a family of highly evolutionary conserved proteins reportedly involved in the regulation of actin cytoskeletal dynamics, although only coronin 3 has been shown to be related to cancer cell migration.
Specifications
Specifications
CORO1C
Polyclonal
Unconjugated
CORO1C
CORO1C, Coronin 1C, Coronin 3, CRNN4, HCRNN4
Rabbit
Antigen Affinity Chromatography
RUO
23603, 23790, 501841
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9ULV4, Q9WUM4
Coro1c
CORO1C Fusion Protein Ag6488
150 μL
Primary
Human, Mouse, Rat