missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 1536-Well, CellBIND-Treated, Flat-Bottom Microplate
GREENER_CHOICE

Product Code. 12721935 Shop All Corning Products
Click to view available options
Color:
Black, Clear Bottom
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 12721935

Brand: Corning™ 3833

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Color:Black, Clear Bottom; Sterility:Sterile; Lid:Without Lid

Corning 1536-well microplates are our highest density microplates available for high throughput screening. The microplates conform to standard microplate footprint and dimensions. These microplates are offered in solid black and white polystyrene, with round or flat bottoms, and in black-clear bottom formats.

  • Total well volume of 10 μL for round well microplates and 12.8 μL for flat bottom microplates
  • Recommended working volume up to 8 μL
  • Round well bottom for reduced air entrapment and improved CV values and Z factor
  • Raised well bottom for higher sensitivity
  • Flood reservoir on four sides to reduce instrument contamination
  • Lids are available separately. Corning lid (Cat. No. 3098) is compatible with these microplates.
TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid Without Lid
Culture Area 0.03 cm2
Array 32 x 48
Color Black, Clear Bottom
For Use With (Equipment) Automated dispensers including the PerkinElmer FlexDrop™
Material Polystyrene
No. of Wells 1536
Volume (Metric) Working 8 μL
Sterility Sterile
Surface Treatment CellBIND
Well Shape Flat Bottom (F-well)
For Use With (Application) For Kalypsys™ systems, optimal bottom reader sensitivity
No. per Case 100/Cs.
No. per Pack 20/Pk.
Quantity 100/Cs.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.