missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Roller Bottle Caps
GREENER_CHOICEBIOPHARMA

Product Code. 10427842 Shop All Corning Products
Change view
Click to view available options
Thread Size (Metric):
31 mm
Unit Size:
Pack of 300
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size (Metric) unitSize
10427842 31 mm Pack of 300
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10427842 Supplier Corning™ Supplier No. 431132
Add to basket
Add to basket
This item is not returnable. View return policy

Easy-grip polyethylene

  • Individually wrapped
  • Sterile

TRUSTED_SUSTAINABILITY

Specifications

Color Red
Material Polyethylene
No. per Case 300/Cs.
No. per Pack 1/Pk.
Sterility Sterile
Thread Size (Metric) 31 mm
Neck Finish 31 mm
For Use With (Equipment) Roller bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.