missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PureCoat™ Collagen I Peptide TC-Treated Microplates
GREENER_CHOICE

Product Code. 15300011
Change view
Click to view available options
Well Count:
6
24
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Well Count unitSize
15300011 6 Pack of 10
15320011 24 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15300011 Supplier Corning™ Supplier No. 356270

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Lid

Coated with biologically active synthetic animal-free peptides that are covalently linked to a proprietary surface

Corning ECM mimetic surfaces and substrates contain biologically active, synthetic, animal-free peptides that have been rationally designed to mimic the cell attachment motifs of native ECM proteins. The peptides enable optimal cell binding and signaling in a broad range of serum-free, xeno-free, and animal-free media formulations, supporting the propagation and differentiation of a range of stem, progenitor, and primary cell types.

Corning PureCoat cultureware is coated with biologically active, synthetic, animal-free peptides that are covalently linked to a proprietary surface to provide a highly consistent, cost-effective alternative to self-coated extracellular peptides. The proprietary covalent linkage orients the peptides for optimal binding and signaling.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance cGMP
Closure Color Clear
Closure Included Yes
Closure Material Polystyrene
Bottom Shape Flat
Color Clear
Culture Area 9.6 cm2
Material Polystyrene
Shape Rectangular
Sterility Sterile
Surface Treatment Cell Culture
Type Cell Culture Microplate
Well Count 6
For Use With (Application) Provides a highly consistent alternative to complex extracellular peptides
Includes Lid
No. per Case 10/Cs.
No. per Pack 5/Pk.
Storage Requirements Room Temperature
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.