Learn More
Corning™ PureCoat™ Carboxyl Surface Dishes
Negatively charged surface improves attachment. increases proliferation and improved differentiation for a broad range of cells that exhibit poor attachment properties in serum-free or serum-reduced conditions
Brand:  Corning™ 356784
Code : 66
Additional Details : Weight : 1.13000kg
 View more versions of this product
Product Code. 11553640
  • £139.39 / Pack of 40
Estimated Shipment: 21-08-2024
to see stock.
Add to basket
Description
Description
Corning PureCoat Amine (positively charged) and Carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.
Specifications
Specifications
Flat
Polystyrene
Carboxyl
40/Cs.
100 mm
58.1 cm2
Round
Cell Culture Dish
10/Pk.