missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ PureCoat™ Amine Flat Bottom Multiwell Plates

Click to view available options
Well Count:
24
Unit Size:
Pack of 5
Description
Corning PureCoat Amine (positively charged) and Carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.
Specifications
Specifications
| Array | 4 x 6 |
| Closure Included | Yes |
| Closure Material | Polystyrene |
| Bottom Shape | Flat |
| Capacity (Metric) | 1 mL (Well) |
| Material | Polystyrene |
| Shape | Rectangular |
| Sterility | Sterile |
| Type | Multi-well Cell Culture Plate |
| Well Count | 24 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction