Learn More
Corning™ PureCoat™ Amine Flat Bottom Multiwell Plates
Description
Corning PureCoat Amine (positively charged) and Carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.
Specifications
Specifications
| Array | 2 x 3 |
| Bottom Shape | Flat |
| Material | Polystyrene |
| Shape | Rectangular |
| Sterility | Sterile |
| Surface Treatment | Amine |
| Type | Multi-well Cell Culture Plate |
| Well Count | 6 |
| For Use With (Application) | Culture of primary cells, transfected, transformed and fastidious cell lines, culture in standard, serum-free or serum-reduced conditions, freeze-thaw recovery of cryopreserved primary cells and cell lines, including division arrested |
| No. per Case | 50/Cs. |
| Show More |