Learn More
Corning™ PureCoat™ Amine Culture Dishes
Description
Corning PureCoat Amine (positively charged) and Carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.
Specifications
Specifications
| Bottom Shape | Flat |
| Culture Area | 58.1 cm2 |
| Material | Polystyrene |
| Shape | Round |
| Surface Treatment | Amine |
| Type | Cell Culture Dish |
| No. per Case | 10/Cs. |
| No. per Pack | 10/Pk. |
| Diameter (Metric) | 100 mm |