missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 384-Well, Poly-D Lysine-Treated, Flat-Bottom Microplate

Product Code. 11399514 Shop All Corning Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11399514 Clear Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11399514 Supplier Corning™ Supplier No. 3844
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Color:Clear; Sterility:Sterile; Lid:With Lid

  • Total well volume of 112 μL; working well volume of 20 μL – 80 μL
  • Cell culture microplates are sterile and nonpyrogenic.
  • The 384-well Universal Optics NBS microplate is manufactured using an advanced polymer with high clarity and improved chemical resistant properties.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Format Standard
    Lid With Lid
    Culture Area 0.06 cm2
    Array 16 x 24
    Color Clear
    Material Polystyrene
    No. of Wells 384
    Volume (Metric) Working 20 to 80 μL
    Sterility Sterile
    Surface Treatment Poly-D-Lysine
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Easy transition into existing automation and readers, reduced evaporation of well contents, enhanced cell attachment
    No. per Case 100/Cs.
    No. per Pack 20/Pk.
    Well Color Clear
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.