Learn More
Corning™ 96-Well, Poly-D Lysine-Treated, Flat-Bottom Microplate
GREENER_CHOICE
Product Code. 11379514 Shop All Corning Products
Change view
Click to view available options
Color:
Black, Clear Bottom
Unit Size:
Pack of 100
Product Code. Color unitSize
11379514 Black, Clear Bottom Pack of 100
1 options
Product Code. 11379514 Supplier Corning™ Supplier No. 3842
  • ONLINE EXCLUSIVE PRICE
    £1990.00
    £1857.22 / Pack of 100
    Save £132.78
    6% Off
In Stock
Add to basket
Add to basket
Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid
  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Opaque walls prevent well-to-well cross-talk.
  • Optically clear flat bottom permits direct microscopic viewing.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Lid With Lid
    Culture Area 0.32 cm2
    Array 8 x 12
    Color Black, Clear Bottom
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 75 to 200 μL
    Sterility Sterile
    Surface Treatment Poly-D-Lysine
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Easy transition into existing automation and readers, reduced evaporation of well contents, enhanced cell attachment, reduced crosstalk and have low background fluorescence and minimal light scatter
    No. per Case 100/Cs.
    No. per Pack 20/Pk.
    Well Color Clear