missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well, Poly-D Lysine-Treated, Flat-Bottom Microplate

Product Code. 11369514 Shop All Corning Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11369514 Clear Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11369514 Supplier Corning™ Supplier No. 3841
  • ONLINE EXCLUSIVE PRICE
    £1225.00
    £1157.81 / Pack of 100
    Save £67.19
    5% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Color:Clear; Sterility:Sterile; Lid:With Lid

  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated

Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.

Cat. Nos. 3585 and 3595 have special low evaporation lids.

Cat. No. 3841 is aseptically manufactured.

TRUSTED_SUSTAINABILITY

Specifications

Lid With Lid
Culture Area 0.32 cm2
Array 8 x 12
Binding Type Poly-D-Lysine
Color Clear
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Sterile
Surface Treatment Poly-D-Lysine
Well Shape Flat Bottom (F-well)
For Use With (Application) Easy transition into existing automation and readers, reduced evaporation of well contents, enhanced cell attachment
No. per Case 100/Cs.
No. per Pack 20/Pk.
Well Color Clear
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.