missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Paddle Assembly for Reusable Spinner Flasks
Shop All Corning Products
Click to view available options
For Use With (Equipment):
15L Corning Reusable spinner flask
36L Corning Reusable spinner flask
8L Corning Reusable spinner flask
Unit Size:
Each
Description
- For series 4510, 4512, and 4514 spinner flasks when coupled to a direct drive motor.
- Paddle assemblies will couple to 100 mm and 120 mm cap assemblies.
Specifications
Specifications
| For Use With (Equipment) | 36L Corning Reusable spinner flask |
| Type | Paddle Assembly |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction