Learn More
Corning™ Osteo Assay Surface Microplates
Capable of supporting the functional properties of osteogenic cells. Corning™ Osteo Assay Surface delivers lot-to-lot consistency, translating to reliable and reproducible results in bone cell assays. The surface also offers a consistent and defined alternative to preparing dentine or bone slices, reducing the variability in assay systems and resulting in more predictable assay readouts.
Brand:  Corning™ 3989
Code : 66
Additional Details : Weight : 0.31751kg
 View more versions of this product
Product Code. 11397791
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Individual alphanumerical codes for well identification, flat bottoms
  • Treated for optimal cell attachment (except where noted)
  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
  • Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
  • Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
  • Sterile
  • Nonpyrogenic
Specifications
Specifications
With Lid
1/Pk.; 2/Cs.
96-well Osteo Assay Surface Multiwell Microplate
360 μL
75 to 200 μL
Osteo Assay
2/Cs.
Clear
8 x 12
Clear
Polystyrene
96
Sterile
Osteoblast and osteoclast differentiation and functional studies
1/Pk.