Learn More
Corning™ Block, 6 x 20 mm Tubes
Product Code. 11373825 Shop All Corning Products
Click to view available options
For Use With (Equipment):
LSE Digital Dry Bath Heater
Unit Size:
Each
Product Code. 11373825
Brand: Corning™ 480127
Only null left
Add to basket
Add to basket
Material: Stainless Steel; For Use With: LSE Digital Dry Bath Heater
The single and dual block configurations are used in the Corning LSE Digital Dry Bath Heaters.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) LSE Digital Dry Bath Heater
Material Stainless Steel
Type Block
For Use With (Application) Holds 6 x 20 mm tubes