missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Low-Volume 384-Well Polystyrene Plates
High-base design is compatible with automation and bar coding
£565.38 - £634.82
Specifications
Format | Standard |
---|---|
Lid | Without Lid |
Array | 16 x 24 |
Material | Polystyrene |
No. of Wells | 384 |
Product Code | Brand | Sterility | Surface Treatment | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sterility | Surface Treatment | Price | Quantity & Availability | ||||
10727033
|
Corning™
3540 |
Non-sterile | Non-treated |
£565.38
Pack of 50 |
Estimated Shipment: 11-09-2024
Log in to see stock. |
||||
10445823
|
Corning™
3544 |
Non-sterile | Non-binding |
£634.82
Pack of 50 |
Estimated Shipment: 11-09-2024
Log in to see stock. |
||||
Description
- Validated for the FLIPR™ TETRA system—outer wells aligned for proper viewing with the FLIPR system
- 2.8mm distance between well bottoms and plate bottom
- Round wells with flat bottoms
- Working volume: 5–45μL
- Untreated surface binds biomolecules through passive interactions; Nonbinding surface (NBS™) creates a nonionic hydrophilic surface that minimizes molecular interactions; Tissue culture-treated surface provides optimal cell attachment
Specifications
Standard | |
16 x 24 | |
384 | |
Non-sterile | |
For miniaturized 384-well fluorescent homogeneous assays |
Without Lid | |
Polystyrene | |
0.005 to 0.04 mL | |
Flat Bottom (F-well) |