missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Low-Volume 384-Well Polystyrene Plates

High-base design is compatible with automation and bar coding

Brand:  Corning™ 3544

Code : 66

Additional Details : Weight : 0.03049kg

 View more versions of this product

Product Code. 10445823

  • £634.82 / Pack of 50
Estimated Shipment: 12-09-2024
to see stock.
Add to basket

Description

Description

  • Validated for the FLIPR™ TETRA system—outer wells aligned for proper viewing with the FLIPR system
  • 2.8mm distance between well bottoms and plate bottom
  • Round wells with flat bottoms
  • Working volume: 5–45μL

  • Untreated surface binds biomolecules through passive interactions; Nonbinding surface (NBS™) creates a nonionic hydrophilic surface that minimizes molecular interactions; Tissue culture-treated surface provides optimal cell attachment

Specifications

Specifications

Standard
16 x 24
Polystyrene
0.005 to 0.04 mL
Non-binding
For miniaturized 384-well fluorescent homogeneous assays
Without Lid
Black
384
Non-sterile
Flat Bottom (F-well)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers