Learn More
Corning™ Fiberglass Prefilters
For use in Corning bottle top and vacuum filtration systems.
Brand:  Corning™ 431411
 View more versions of this product
Product Code. 11353775
  • £73.38 / Pack of 100
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For use with vacuum filtration systems or bottle top vacuum filters
TRUSTED_SUSTAINABILITY
Specifications
Specifications
250 mL filter funnels, Corning bottle top and vacuum filtration systems
Non-sterile
100/Pk.
Fiberglass
100
Prefilter