Learn More
Falcon™ 96-well Feeder Tray and Lid
GREENER_CHOICE
Product Code. 10210822 Shop All Corning Products
Change view
Click to view available options
No. of Wells:
96
Unit Size:
Pack of 5
Product Code. No. of Wells unitSize
10210822 96 Pack of 5
1 options
Product Code. 10210822 Supplier Falcon™ Supplier No. 353924
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ideal for both manual and robotic screening of compounds in cell-based assays
Falcon™ 96-well Feeder Tray, with lid. Non-TC-treated Polystyrene cell media tray for feeding cell-based assays. The drop-in baffle for the Feeder Tray mitigates media sloshing and lowers the risk of contamination.
  • Gamma irradiated.
  • Nonpyrogenic.
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid With Lid
Array 8 x 12
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Non-treated
No. per Case 5/Cs.
No. per Pack 5/Pk.
Quantity 5/Cs.