Learn More
Corning™ Extracellular Matrix, Human
Product Code. 10021021 Shop All Corning Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. Quantity unitSize
10021021 1 mg 1mg
1 options
Product Code. 10021021 Supplier Corning™ Supplier No. 354237
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A chromatographically partially purified matrix extract derived from human placenta
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Extracellular matrix
Quantity 1 mg
Content And Storage -70°C