missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Ready to use without surface preparation, prehybridization, or addition of expensive crosslinking reagents or highly purified oligos
- Ideal for hybridization and amplification
- Surface provides binding with high specificity and extremely low background
- Flat-bottom wells
Specifications
Specifications
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Color | Clear |
| Material | Polystyrene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Well Shape | Round |
| For Use With (Application) | Ideal for hybridization and amplification |
| No. per Case | 10/Cs. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
