Learn More
Description
Feature a constriction one inch below the top to facilitate plugging. Corning™ Disposable Glass Pasteur Pipets are ideal for nonvolumetric transfer work in bacteriology, immunology, hematology and serology studies.
Corning disposable glass Pasteur pipets are available in convenient Packages. One-time use eliminates the danger of cross-contamination of valuable specimens or laboratory reagents. These pipets feature a constriction one inch below the top to facilitate plugging. They are ideal for rapid nonvolumetric transfer work in bacteriology, immunology, hematology, and serology studies, as well as blood bank and clinical chemistry procedures.
'B' series pipets are soda lime glass. 'D' series are borosilicate glass.
Specifications
Specifications
| Aspirator Plug | No |
| Diameter (English) Outer | 0.255 in. |
| Diameter (Metric) Outer | 0.65 cm |
| Material | Soda Lime Glass |
| Sterility | Non-sterile |
| Includes | 1 in. Constriction below the top |
| Length (English) | 5.75 in. |
| Tip Style | Round |
| Length (Metric) | 14.6 cm |
| Type | Unplugged Pasteur Pipet |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.